ContactUsLinkComponent
Stericlin® container seal, loop into container lock and fold to seal, both halves blank, no indicator or documentation field, 80x40m m (40x40mm folded), strip 2.5x70mm 1 * 500 items
  VEPA3FCZB460104
Stericlin® container seal, loop into container lock and fold to seal, both halves blank, no indicator or documentation field, 80x40m m (40x40mm folded), strip 2.5x70mm 1 * 500 items
  VEPA3FCZB460104
Frequently Bought Together