ContactUsLinkComponent
Agilent J+W HP-5 Intuvo GC column module, 25 m, 0.20 mm, 0.5 µm 1 * 1 items
Catalog # HEWL19091J-202-INT
Supplier:  Agilent
Agilent J+W HP-5 Intuvo GC column module, 25 m, 0.20 mm, 0.5 µm 1 * 1 items
Catalog # HEWL19091J-202-INT
Supplier:  Agilent
Frequently Bought Together