ContactUsLinkComponent
Agilent J+W HP-FFAP Intuvo GC column module, 50 m, 0.20 mm, 0.33 µm 1 * 1 items
Catalog # HEWL19091F-105-INT
Supplier:  Agilent
Agilent J+W HP-FFAP Intuvo GC column module, 50 m, 0.20 mm, 0.33 µm 1 * 1 items
Catalog # HEWL19091F-105-INT
Supplier:  Agilent
Frequently Bought Together