Order Entry
Austria
Orders LinkContactUsLinkComponent
Mop systems, Edgeless®

Mop systems, Edgeless®

Edgeless floor mop frame QuickConnect™, polypropylene copolymer frame with stainless steel connector ; compatible with all styles of Edgeless mops and easily attaches to QuickConnect handles. Autoclavable at 121°C for 30 min., 150×180 mm, -, -
Catalog # CONT2646F
Supplier:   CONTEC
Mop systems, Edgeless®
Catalog # CONT2646F
Supplier:  CONTEC
Mop systems, Edgeless®
Mop systems, Edgeless®

Edgeless floor mop frame QuickConnect™, polypropylene copolymer frame with stainless steel connector ; compatible with all styles of Edgeless mops and easily attaches to QuickConnect handles. Autoclavable at 121°C for 30 min., 150×180 mm, -, -

Price
193,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Designed for wet mopping in controlled environments and cleanrooms for effective cleaning and retrieval of excess fluid. Ideal for use in sterile environments. Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption.

  • Mop heads are free from any cellulose contaminants
  • Stainless bucket, downpress wringer and insert maximise wringing pressure and effectiveness
  • All system components and buckets are compatible with gamma, ETO, and autoclave sterilisation

Mop heads are laundered and packaged in a ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.

Ordering information: All mop system components must be ordered separately. Plastic bucket systems include cart, buckets, wringer, and casters. Stainless steel bucket systems include trolley, buckets, wringer, and casters.

Specifications
  • Capacity:
    -
  • Description:
    Edgeless floor mop frame QuickConnect™, polypropylene copolymer frame with stainless steel connector ; compatible with all styles of Edgeless mops and easily attaches to QuickConnect handles. Autoclavable at 121°C for 30 min.
  • Dimensions:
    150×180 mm
  • Length:
    -
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.