Order Entry
Austria
Orders LinkContactUsLinkComponent
WB blocking buffer kit, TrueBlack®

WB blocking buffer kit, TrueBlack®

TrueBlack® WB blocking buffer kit, 10 membranes, 1 KIT
Catalog # 23013-T
Supplier:   Biotium
WB blocking buffer kit, TrueBlack®
Catalog # 23013-T
Supplier:  Biotium
CAS Number:  
WB blocking buffer kit, TrueBlack®

TrueBlack® WB blocking buffer kit, 10 membranes, 1 KIT

CAS Number:  
Price
133,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

The TrueBlack® WB blocking buffer kit is a ready-to-use buffer system that provides optimal specificity, sensitivity, and background signal suppression for fluorescence-based western blotting (WB).

  • Blocks non-specific protein binding, reducing background fluorescence over the entire membrane
  • Suppresses background caused by charged fluorescent dyes (performs better than BSA, gelatin, or casein)
  • Reduces antibody cross-reactivity, eliminating non-specific bands
  • Compatible with both polyvinylidene difluoride (PVDF) and nitrocellulose membranes
  • Can be used with antibodies labeled with all types of fluorophores spanning the visible and NIR spectrum
  • Contains non-mammalian blocking agents, for broad secondary antibody compatibility
  • TBS based buffer suitable for phosphoprotein detection

The buffers are designed to achieve optimal specificity and sensitivity by blocking background from the non-specific interaction between the fluorophore-labeled antibodies and the membrane. Non-specific signal in WB can arise from multiple sources, including antibody cross-reactivity with off-target proteins, non-specific antibody adsorption to the membrane, and membrane autofluorescence. Another potential cause of background that is not well-known is the effect of fluorescent dyes themselves on the specificity of labeled antibodies.

Next-generation fluorescent dyes like Alexa Fluor® or CF® dyes often carry multiple negative charges to improve dye solubility and brightness of conjugates. The extra charge carried by the dye can result in non-specific antibody binding and background fluorescence. The TrueBlack® WB Blocking Buffer Kit blocks background from multiple sources including charged dye conjugates. It is especially advantageous for phosphoprotein detection, significantly improving specificity compared to conventional blocking buffers.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    TrueBlack® WB blocking buffer kit, 10 membranes

Specifications