Thermo Scientific Capitol Vial containers with patented flip-top integrally hinged to the container. Food Grade Polypropylene Aseptic-By-Process. Capacity 2,5 oz (75 mL). ID 50 mm. Height 50 mm. 1 * 400 items
Catalog # SAMC2.5CL
Supplier: SAMCO
Frequently Bought Together