tri-Sodium citrate dihydrate EMPROVE ESSENTIAL Ph Eur,BP,JP,USP,E 331 1 * 1 kg
Catalog # 1.06447.1000
Supplier: MERCK PRODUCTION CHEMICALS
tri-Sodium citrate dihydrate EMPROVE ESSENTIAL Ph Eur,BP,JP,USP,E 331 1 * 1 kg
Catalog # 1.06447.1000
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Reagent reservoirs
Supplier: Brand
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Eppendorf® Pipette Holder System
Supplier: EPPENDORF