Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Microplate reader, single-mode, SpectraMax® ABS

SpectraMax® ABS microplate reader, single-mode, with current version of SoftMax® Pro and 2 additional years MD service plan covering parts and labour
Catalog # MLDVABS-ACAD3
Supplier:   Molecular Devices
Microplate reader, single-mode, SpectraMax® ABS
Catalog # MLDVABS-ACAD3
Supplier:  Molecular Devices
Microplate reader, single-mode, SpectraMax® ABS
Microplate reader, single-mode, SpectraMax® ABS

SpectraMax® ABS microplate reader, single-mode, with current version of SoftMax® Pro and 2 additional years MD service plan covering parts and labour

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

SpectraMax® ABS microplate reader bridges the gap between the affordability of filter-based readers and the flexibility of monochromator-based systems. Equipped with advanced technology that provides flexibility, sensitivity, and convenience for a wide range of assays such as ELISA, microbial growth, and protein quantitation, all in an incredibly small footprint.

  • Compact design
  • Minimum volume and maximum throughput with 96-well microplate compatibility
  • Powerful data analysis with SoftMax® Pro Software
  • Optimal performance with validation tools

SpectraMax® ABS readers use a grating monochromator to select the exact wavelength needed for every assay. The ABS reader ranges from 340 to 850 nm in wavelength. The advanced optical and electronic design of the readers give the same high performance with round-bottom, flat-bottom or half-area well plates. Select upto six wavelengths in a single read to maximise the number of data points collected in an experiment.

Specifications
  • Description:
    SpectraMax® ABS microplate reader, single-mode, with current version of SoftMax® Pro and 2 additional years MD service plan covering parts and labour
  • Seq:
    03
  • Incubation range:
    5 °C above ambient to 45 °C
  • Light source:
    Xenon flash lamp
  • Temperature uniformity:
    ±0,5 °C
  • Photometric range:
    0,000 - 4,000 OD
  • Shaking:
    Linear
  • Wavelength repeatability:
    ±0,2 nm
  • Wavelength bandwidth:
    2,0 nm
  • Photometric accuracy:
    <±0,006 OD ±1%
  • Wavelength accuracy:
    ±1 nm
  • Weight:
    11,3 kg
  • Cat. No.:
    MLDVABS-ACAD3
  • Wavelength selection:
    Monochromator, tunable in 1,0 nm increments
  • Photometric linearity:
    0,0 - 3,0 OD
  • Stray light:
    <0,05 % at 230 nm
  • Model:
    SpectraMax® ABS
  • Wavelength range:
    340 - 850 nm
  • W×D×H:
    305×414×203 mm
  • Plate types:
    96-well plates
  • Software:
    SoftMax® Pro
  • Measurement time:
    9 s in normal read mode, 5 s in speed read mode
  • Measuring modes:
    End-point and kinetics