ContactUsLinkComponent
Stericlin® sterilisation wrap sheet, SSMMS 43g/m², one side light blue, one side violet, 90x90cm, DIN EN ISO 11607 1 * 200 items
Catalog # 129-1361
Stericlin® sterilisation wrap sheet, SSMMS 43g/m², one side light blue, one side violet, 90x90cm, DIN EN ISO 11607 1 * 200 items
Catalog # 129-1361
Frequently Bought Together