Orders LinkContactUsLinkComponent
Tool carts
Tool carts
Tool cart, with 5 drawers, secure lock and cleanroom castors with brakes, 730×470×850 mm
Catalog # PALBCT0702
Supplier:   Palbam
Tool carts
Catalog # PALBCT0702
Supplier:  Palbam
Tool carts
Tool carts
Tool cart, with 5 drawers, secure lock and cleanroom castors with brakes, 730×470×850 mm
Price
7 020,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Tool cart in stainless steel 304 with electropolished finish.
  • Five fully extendable drawers
  • One lock which secures all the drawers
  • Top ledge to prevent tools/parts falling
  • Cleanroom castors with brakes
Specifications
  • Description:
    Tool cart, with 5 drawers, secure lock and cleanroom castors with brakes
  • Seq:
    1
  • W×D×H:
    730×470×850 mm