ContactUsLinkComponent
2002-9125 - 4ml HDPE bottle (with 13mm finish cap processed separately) Process: Particulate Cleaning & Gamma Irradiation. Packaging: Single cleanroom bag each bottle. Certification: C of A Endotoxin (Report Actual Results), C of A USP Particles, C o 1 * 72 items
Catalog # NALGCEE7M4PI-EP
2002-9125 - 4ml HDPE bottle (with 13mm finish cap processed separately) Process: Particulate Cleaning & Gamma Irradiation. Packaging: Single cleanroom bag each bottle. Certification: C of A Endotoxin (Report Actual Results), C of A USP Particles, C o 1 * 72 items
Catalog # NALGCEE7M4PI-EP
Frequently Bought Together