Order Entry
Austria
Orders LinkContactUsLinkComponent
Mouse Recombinant FGF-Basic (from <i>E. coli</i>)

Mouse Recombinant FGF-Basic (from E. coli)

Vial, 50 µG
Catalog # SHBT200-12-50UG
Mouse Recombinant FGF-Basic (from <i>E. coli</i>)
Catalog # SHBT200-12-50UG
CAS Number:  
Mouse Recombinant FGF-Basic (from <i>E. coli</i>)
Mouse Recombinant FGF-Basic (from E. coli)

Vial, 50 µG

CAS Number:  
Price
190,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. The application of FGF-basic is a critical component for human embryonic stem cell culture systems and is necessary for maintaining human embryonic stem cells in an undifferentiated state.

  • High quality
  • Low endotoxin
  • Affordable

Caution: For research use only.

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    50 µG
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    16,5 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Protein function:
    Cytokine
  • Gene ID:
    P15655
  • Formulation:
    10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
  • Endotoxin-free:
    N
  • Tested applications:
    Bioassay
  • Species:
    Mouse
  • Protein/peptide name:
    FGF-Basic
  • Protein synonyms:
    Prostatropin|HBGF-2|FGF2
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • Biological activity:
    Bioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mg
  • Shipping temperature:
    RT
  • Cat. No.:
    SHBT200-12-50UG
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Storage conditions:
    –20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    N

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.