Order Entry
Austria
Orders LinkContactUsLinkComponent
Rat Recombinant FGF-basic (from <i>E. coli</i>)

Rat Recombinant FGF-basic (from E. coli)

Vial, 100 µG
Catalog # SHBT300-02-100UG
Rat Recombinant FGF-basic (from <i>E. coli</i>)
Catalog # SHBT300-02-100UG
CAS Number:  
Rat Recombinant FGF-basic (from <i>E. coli</i>)
Rat Recombinant FGF-basic (from E. coli)

Vial, 100 µG

CAS Number:  
Price
344,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic binds heparin in order to signal through fibroblast growth factor (FGF) receptor tyrosine kinases.

  • High Quality
  • Low Endotoxin
  • Affordable

Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.

Caution: For research use only.

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    100 µG
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    16,4 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Protein function:
    Cytokine
  • Gene ID:
    P13109
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • Endotoxin-free:
    N
  • Tested applications:
    Bioassay
  • Species:
    Rat
  • Protein/peptide name:
    FGF-basic
  • Protein synonyms:
    Prostatropin|HBGF-2|FGF2
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • Biological activity:
    Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
  • Shipping temperature:
    RT
  • Cat. No.:
    SHBT300-02-100UG
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Storage conditions:
    –20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    N

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.