Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat; 0,1 NA, 18,5 mm
Catalog # ECHLJMAG0044
Supplier:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Catalog # ECHLJMAG0044
Supplier:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat; 0,1 NA, 18,5 mm
Price
118,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
  • Description:
    Plan achromat; 0,1 NA
  • Seq:
    04
  • Working distance:
    18,5 mm