Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite phase objective, 17 mm
Catalog # 630-3052
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # 630-3052
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Fluorite phase objective, 17 mm
Price
831,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
  • Description:
    Fluorite phase objective
  • Seq:
    29
  • Working distance:
    17 mm