Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Apochromat objective, 5,0 mm
Catalog # ECHLJMAG0011
Supplier:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Catalog # ECHLJMAG0011
Supplier:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Apochromat objective, 5,0 mm
Price
2 270,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    1,25×
  • Description:
    Apochromat objective
  • Seq:
    01
  • Working distance:
    5,0 mm