Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat oil phase objective, 0,15 mm
Artikelnr. ECHLJMAG1006
Anbieter:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Artikelnr. ECHLJMAG1006
Anbieter:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat oil phase objective, 0,15 mm
Preis
984,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    100×
  • Description:
    Achromat oil phase objective
  • Seq:
    27
  • Working distance:
    0,15 mm