Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat phase objective, 1,2 mm
Catalog # ECHLJMAG0207
Supplier:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Catalog # ECHLJMAG0207
Supplier:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat phase objective, 1,2 mm
Price
505,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    20×
  • Description:
    Achromat phase objective
  • Seq:
    13
  • Working distance:
    1,2 mm