Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
Fluorite oil phase objective, 0,2 mm
Catalog # 630-3068
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # 630-3068
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Fluorite oil phase objective, 0,2 mm
Price
4 000,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    100×
  • Description:
    Fluorite oil phase objective
  • Seq:
    08
  • Working distance:
    0,2 mm