Colour coded inserts, CapInsert™
Red
SIMPM957R
These colour coded inserts could be applied to the tube closure, which eliminates jeopardising the integrity of your sample by transferring it to another vial.
- Manually press fit and locks into place on top of closure
- Colour:Red
- Seq:04
- Cat. No.:SIMPM957R