Orders LinkContactUsLinkComponent
Colour coded inserts, CapInsert™
Colour coded inserts, CapInsert™
Yellow
Catalog # SIMPM957Y
Supplier:   Simport Scientific
Colour coded inserts, CapInsert™
Catalog # SIMPM957Y
Supplier:  Simport Scientific
Colour coded inserts, CapInsert™
Colour coded inserts, CapInsert™
Yellow
Price
28,20 €
Log in for availability
1 * 200 ST
Product Details & Documents
These colour coded inserts could be applied to the tube closure, which eliminates jeopardising the integrity of your sample by transferring it to another vial.
  • Manually press fit and locks into place on top of closure
Specifications
  • Colour:
    Yellow
  • Seq:
    05