Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Ratiolab® Cryo-Boxes, PP, grid 9 x 9, green, 133 x 133 x 50/75 mm, combi-lid, numerically coded 1 * 5 items
Catalog # RATI5320013
Supplier:
RATIOLAB
Expand image
Ratiolab® Cryo-Boxes, PP, grid 9 x 9, green, 133 x 133 x 50/75 mm, combi-lid, numerically coded 1 * 5 items
Catalog # RATI5320013
Supplier:
RATIOLAB
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
Repetitive pipette, Multipette® E3 / E3x
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection