BUFFER SOLUTION PH12.00 +0.05 25GRAD 1 * 500 mL
Catalog # REGE1120525TT
Supplier: REAGECON
Frequently Bought Together
Analytical Volumetric Solution Zinc Sulphate 0.1N 0.1M 1 * 1 L
Supplier: REAGECON
Supelco® PESTANAL® Organic Reference Standard, Hexazinone
Supplier: Merck
TRIGLYCINE 1 * 100 mg
Supplier: Merck
BUFFER SOLUTION PH1.00+0.02 25GRAD 1 * 500 mL
Supplier: REAGECON
Methyl Linolenate, Reference Standard
Supplier: USP
PAH-Mix 9 deuterated 100 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
Buffer solutions
Supplier: REAGECON
Methacrylamide
Supplier: EHRENSTORFER
Pesticide reference standard, cis-Permethrin 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Supplier: EHRENSTORFER
Tantalum standard solution, for ICP, in 2% HF, CRM, VHG
Supplier: LGC Standards PROMOCHEM
SOYASAPOGENOL B 1 * 5 mg
Supplier: Merck
AMIODARONE RELATED COMPOUND E 1 * 100 mg
Supplier: Merck
Buffer PH 4,00 1 * 500 mL
Supplier: REAGECON
Vardenafil Related Compounds, Reference Standard
Supplier: USP