BUFFER SOLUTION PH1.00+0.02 25GRAD 1 * 500 mL
Catalog # REGE1010525TT
Supplier: REAGECON
Frequently Bought Together
Syringe filters, Acrodisc®, PTFE membrane
Supplier: Cytiva (Formerly Pall Lab)
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions
Supplier: REAGECON
BUFFER PH 7,0 +/- 0,01 SOLUTION 25 1 * 500 mL
Supplier: REAGECON