Storage Boxes for Vials
Storage box for EPA screw neck vials N 24 (20 ml, 30 ml, 40 m), 16, 130×130×102 mm
Catalog # MANA702517
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Boxes are made from blue plastic with integrated or removable divider and transparent lid.
- Allow a secure transportation of sample vials
- Safe standing position in dividers designed for the respective diameter
- Available for all 1,5 ml vials (standard volume), for crimp and screw neck vials N 13 and for headspace vials with screw neck N 18 or crimp neck N 20, respectively
- Also available for EPA screw neck vials N 24 as well as for storage vials N 15
Ideal for space-saving storage in fridges and freezers, since the transparent lid prevents condensations on the closures and thus avoids a possible contamination in the cooling unit.
Dokumente
Vials and Caps.pdf
Weitere Produktdetails
Specifications
- Compartments:16
- W×D×H ext.:130×130×102 mm
- Description:Storage box for EPA screw neck vials N 24 (20 ml, 30 ml, 40 m)
- Seq:04
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.



