ContactUsLinkComponent
NucleoBond Buffer Set III for the isolation of genomic DNA from bacteria using NucleoBond AXG columns, sufficient for 10 AXG 500 col umns (Maxi preps), 20 AXG 100 columns (Midi preps), or 100 AXG 20 columns (Mini preps) 1 * 1 SET
Catalog # MANA740603
Supplier:  MACHEREY-NAGEL
undefined
NucleoBond Buffer Set III for the isolation of genomic DNA from bacteria using NucleoBond AXG columns, sufficient for 10 AXG 500 col umns (Maxi preps), 20 AXG 100 columns (Midi preps), or 100 AXG 20 columns (Mini preps) 1 * 1 SET
Catalog # MANA740603
Supplier:  MACHEREY-NAGEL
Frequently Bought Together