ContactUsLinkComponent
Impact rotor with 6 ribs and cooling fins, pure titanium, fast comminution of fibrous substances - feed size < 15 mm null
Catalog # FRIT14.4430.32
Supplier:  FRITSCH
Impact rotor with 6 ribs and cooling fins, pure titanium, fast comminution of fibrous substances - feed size < 15 mm null
Catalog # FRIT14.4430.32
Supplier:  FRITSCH
Frequently Bought Together