ContactUsLinkComponent
Cartridge with cpl. dry measuring cell incl. insertion block - for the use as a conversion kit - for insertion in the dry dispersio n unit (article no. 22.8600.00) for using the dry dispersion unit as falling chute or for insertion in the falling ch 1 * 1 items
Catalog # FRIT22.8035.00
Supplier:  FRITSCH
Cartridge with cpl. dry measuring cell incl. insertion block - for the use as a conversion kit - for insertion in the dry dispersio n unit (article no. 22.8600.00) for using the dry dispersion unit as falling chute or for insertion in the falling ch 1 * 1 items
Catalog # FRIT22.8035.00
Supplier:  FRITSCH
Frequently Bought Together