Sieve cassette 4 mm square perforation, made of chromium-free steel ST12 for Power Cutting Mill PULVERISETTE 25 null
FRIT45.7500.09
: FRITSCH
Sieve cassette 4 mm square perforation, made of chromium-free steel ST12 for Power Cutting Mill PULVERISETTE 25 null
FRIT45.7500.09
: FRITSCH
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection