Orders LinkContactUsLinkComponent
Screw caps
Screw caps
50 mm, 500/750/1000 ml
Catalog # 215-4245
Supplier:   Kautex
Screw caps
Catalog # 215-4245
Supplier:  Kautex
Screw caps
Screw caps
50 mm, 500/750/1000 ml
Price
90,60 €
Log in for availability
1 * 100 ST
Product Details & Documents
LDPE, natural.
  • Good chemical resistance
  • Good thermal resistance
  • Leakproof
Specifications
  • Thread:
    50 mm
  • Seq:
    05
  • For bottles:
    500/750/1000 ml