ContactUsLinkComponent
elma tec clean A3 Cleans surfaces made of stainless steel, steel and other iron-metals, titanium and precious metals after mechanical treatment (turning, milling, lapping, polishing, grinding, drawing and punching). 1 * 25 L
Catalog # ELMA8000120
Supplier:  ELMA SCHMIDBAUER
elma tec clean A3 Cleans surfaces made of stainless steel, steel and other iron-metals, titanium and precious metals after mechanical treatment (turning, milling, lapping, polishing, grinding, drawing and punching). 1 * 25 L
Catalog # ELMA8000120
Supplier:  ELMA SCHMIDBAUER
Frequently Bought Together