Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR® Basic Mob Caps, Single Elastic

VWR® Basic Mob Caps, Single Elastic

Blue, 530 mm
Catalog # 113-8851
Supplier:   VWR Collection
VWR® Basic Mob Caps, Single Elastic
Catalog # 113-8851
Supplier:  VWR Collection
VWR® Basic Mob Caps, Single Elastic
VWR® Basic Mob Caps, Single Elastic

Blue, 530 mm

Price
58,50 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

These disposable pleated caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.

  • Free from natural rubber latex
  • Single elastic
  • Available in a wide range of colours

Weight: 10 g/m²

Verpackung: 100 pieces per bag, 10 bags per case.

Specifications
  • Ø:
    530 mm
  • Colour:
    Blue
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.