ContactUsLinkComponent
5 ml Precision FAST qPCR Master Mix with ROX premixed with SYBRgreen (5x 1ml) 1 * 500 Tests
Catalog # ZPFASTRSY5ML
Supplier:  PRIMERDESIGN
5 ml Precision FAST qPCR Master Mix with ROX premixed with SYBRgreen (5x 1ml) 1 * 500 Tests
Catalog # ZPFASTRSY5ML
Supplier:  PRIMERDESIGN
Frequently Bought Together