ContactUsLinkComponent
1 ml Precision FAST qPCR Master Mix for capillary LightCycler 1.0-2.0 premixed with SYBRgreen 1 * 100 Tests
  ZPFASTCLSY1ML
1 ml Precision FAST qPCR Master Mix for capillary LightCycler 1.0-2.0 premixed with SYBRgreen 1 * 100 Tests
  ZPFASTCLSY1ML
Frequently Bought Together