SILICON PLUG FOR ACCESS PORT 1 * 1 items
Catalog # VWRISRV_136
Supplier: VWR Collection
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Accessories for accu-jet® pro
Supplier: Brand
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Accessories for PIPETBOY acu / pro
Supplier: Integra
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
Grade 113V qualitative filter papers, fluted, Whatman™
Supplier: Whatman products (Cytiva)
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG