ContactUsLinkComponent
Breather bag Precision Clean, 1 side Tyvek 1059B, 1 side clear High Density PolyEthylene, without print or sterilisation indicators, suitable for gamma, ethylene oxide or steam sterilisation, suitable for use in cleanrooms, manufactured and packed in 1 * 300 items
Catalog # FISH90728P
Supplier:  FISHER CONTAINER
Breather bag Precision Clean, 1 side Tyvek 1059B, 1 side clear High Density PolyEthylene, without print or sterilisation indicators, suitable for gamma, ethylene oxide or steam sterilisation, suitable for use in cleanrooms, manufactured and packed in 1 * 300 items
Catalog # FISH90728P
Supplier:  FISHER CONTAINER
Frequently Bought Together