ContactUsLinkComponent
Tubing Precision Clean II, made of clear amine-free Low Density PolyEthylene, suitable for use in cleanrooms, manufactured and packed in an ISO 4 cleanroom, thickness 4mil (100 micron) per layer, width/opening 48 (1219mm), length 500feet (152m), 1 r 1 * 1 Roll
Catalog # FISH1N400480
Supplier:  FISHER CONTAINER
Tubing Precision Clean II, made of clear amine-free Low Density PolyEthylene, suitable for use in cleanrooms, manufactured and packed in an ISO 4 cleanroom, thickness 4mil (100 micron) per layer, width/opening 48 (1219mm), length 500feet (152m), 1 r 1 * 1 Roll
Catalog # FISH1N400480
Supplier:  FISHER CONTAINER
Frequently Bought Together