HANDLE OF LOCKING PRECELLYS EVOLUTION 1 * 1 items
Catalog # BERTS000822-PEVO0A
Supplier: Bertin Instruments
HANDLE OF LOCKING PRECELLYS EVOLUTION 1 * 1 items
Catalog # BERTS000822-PEVO0A
Supplier: Bertin Instruments
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
VWR® Aquaresist Water Bath Protective Agent
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG