CONTAINMENT SEAL FOR PRECELLYS EVOLUTION 1 * 1 items
Catalog # BERTS000805-PEVO0A
Supplier: Bertin Instruments
CONTAINMENT SEAL FOR PRECELLYS EVOLUTION 1 * 1 items
Catalog # BERTS000805-PEVO0A
Supplier: Bertin Instruments
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Accessories for PIPETBOY acu / pro
Supplier: Integra
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Repetitive pipette, Multipette® E3 / E3x
Supplier: EPPENDORF