ContactUsLinkComponent
SEAL CONTAINMENT PRECELLYS Sparepart for Precellys 24 and Precellys Dual 24 - seal between mechanical and biologic aera Recommanded frequency placement : 1 year This spare parts can be replaced by the user as described in the user manual 1 * 1 items
Catalog # 432-0241
Supplier:  Bertin Instruments
SEAL CONTAINMENT PRECELLYS Sparepart for Precellys 24 and Precellys Dual 24 - seal between mechanical and biologic aera Recommanded frequency placement : 1 year This spare parts can be replaced by the user as described in the user manual 1 * 1 items
Catalog # 432-0241
Supplier:  Bertin Instruments
Frequently Bought Together