Shipping system insulated ChillTherm regional 17L 2-8C provides temperature control for up to 84h 1 * 6 items
SCATCTR17
Shipping system insulated ChillTherm regional 17L 2-8C provides temperature control for up to 84h 1 * 6 items
SCATCTR17
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection