ContactUsLinkComponent
Precision thermometer made of Quartz-Glass, -10+650:2°C, 450x8-9mm, enclosed scale, cap. colourless, gallium filled, pin-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARQ146032-GA
Supplier:  Amarell
Precision thermometer made of Quartz-Glass, -10+650:2°C, 450x8-9mm, enclosed scale, cap. colourless, gallium filled, pin-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARQ146032-GA
Supplier:  Amarell
Frequently Bought Together