ContactUsLinkComponent
Precision thermometer made of Quartz-Glass, +100+360:0,5°C, 400x8-9mm, enclosed scale, cap. colourless, gallium filled, pin-top fini sh with button, suitable for government verification 1 * 1 items
Catalog # AMARQ146012-GA
Supplier:  Amarell
Precision thermometer made of Quartz-Glass, +100+360:0,5°C, 400x8-9mm, enclosed scale, cap. colourless, gallium filled, pin-top fini sh with button, suitable for government verification 1 * 1 items
Catalog # AMARQ146012-GA
Supplier:  Amarell
Frequently Bought Together