ContactUsLinkComponent
Quartz-Glass therm., +400+800:2°C, upper part c. 300x13-15mm, lower part 600x8-9mm, milk-glass scale, cap. colourless, gallium fille d, partial immersion is by 5-10cm shorter than the stem, cork-plaster-top finish, with metal cap, suitable for govern 1 * 1 items
Catalog # AMARQ137140-GA
Supplier:  Amarell
Quartz-Glass therm., +400+800:2°C, upper part c. 300x13-15mm, lower part 600x8-9mm, milk-glass scale, cap. colourless, gallium fille d, partial immersion is by 5-10cm shorter than the stem, cork-plaster-top finish, with metal cap, suitable for govern 1 * 1 items
Catalog # AMARQ137140-GA
Supplier:  Amarell
Frequently Bought Together