ContactUsLinkComponent
Therm. to set Amarell Universal, +300+700:2°C, 400x9mm, enclosed scale, cap. round, colourless, gallium filled, total immersion, pin -top finish 1 * 1 items
Catalog # AMARN71024-GA
Supplier:  Amarell
Therm. to set Amarell Universal, +300+700:2°C, 400x9mm, enclosed scale, cap. round, colourless, gallium filled, total immersion, pin -top finish 1 * 1 items
Catalog # AMARN71024-GA
Supplier:  Amarell
Frequently Bought Together