ContactUsLinkComponent
Therm. to set Amarell Universal, -1+52:0,1C, 400x9mm, enclosed scale, capillary tube specially coated inside, with non-wetting blu e special liquid, total immersion, richter-top finish, null
Catalog # AMARN71006-CB/WC
Supplier:  Amarell
Therm. to set Amarell Universal, -1+52:0,1C, 400x9mm, enclosed scale, capillary tube specially coated inside, with non-wetting blu e special liquid, total immersion, richter-top finish, null
Catalog # AMARN71006-CB/WC
Supplier:  Amarell
Frequently Bought Together