ContactUsLinkComponent
Therm. to set Amarell Universal, -58+5:0,1C, 400x9mm, enclosed scale, cap.lens front, colourless, toluene filled, total immersion, richter-top finish, null
Catalog # AMARN71004-TOL/WC
Supplier:  Amarell
Therm. to set Amarell Universal, -58+5:0,1C, 400x9mm, enclosed scale, cap.lens front, colourless, toluene filled, total immersion, richter-top finish, null
Catalog # AMARN71004-TOL/WC
Supplier:  Amarell
Frequently Bought Together