API Gravity hydrometer ASTM, 2H-62, 9+21:0,1API, ref. temp. 60F, without therm., 330mm long 1 * 1 items
AMARH825002
: Amarell
API Gravity hydrometer ASTM, 2H-62, 9+21:0,1API, ref. temp. 60F, without therm., 330mm long 1 * 1 items
AMARH825002
: Amarell
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection