ContactUsLinkComponent
API Gravity thermo- hydrometer ASTM, S557HL-14, 59+71:0,1API, designation L, with therm. 0+150:2F with red or blue special liquid, ref. temp. 60F, c.380mm long 1 * 1 items
Catalog # AMARH825382-FL
Supplier:  Amarell
API Gravity thermo- hydrometer ASTM, S557HL-14, 59+71:0,1API, designation L, with therm. 0+150:2F with red or blue special liquid, ref. temp. 60F, c.380mm long 1 * 1 items
Catalog # AMARH825382-FL
Supplier:  Amarell
Frequently Bought Together