ContactUsLinkComponent
Thermometer similar to IP 98 C, solid stem, white backed, 100+300:2°C, gallium filled, durable pigment, suitable for government veri fication 1 * 1 items
Catalog # AMARA341390-GA
Supplier:  Amarell
Thermometer similar to IP 98 C, solid stem, white backed, 100+300:2°C, gallium filled, durable pigment, suitable for government veri fication 1 * 1 items
Catalog # AMARA341390-GA
Supplier:  Amarell
Frequently Bought Together